Assay Details
| Gene Symbol: DHRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010299.4 | XP_004010348.1 | 8 - 9 | 1083 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DHRS1| Gene Symbol: DHRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010299.4 | XP_004010348.1 | 8 - 9 | 1083 | 62 | |
Set Membership: |
|