Assay Details
| Gene Symbol: ASPHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020891.4 | XP_004020940.1 | 2 - 3 | 1678 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASPHD1| Gene Symbol: ASPHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020891.4 | XP_004020940.1 | 2 - 3 | 1678 | 64 | |
Set Membership: |
|