Assay Details
| Gene Symbol: ARID5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021392.4 | XP_004021441.1 | 5 - 6 | 935 | 71 | |
| XM_004021393.4 | XP_004021442.1 | 2 - 3 | 511 | 71 | ||
More Information
Set Membership: |
|

ARID5B
Genomic Map