Assay Details
| Gene Symbol: CHPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094829.2 | XP_014950315.2 | 8 - 9 | 1194 | 87 | |
| XM_027967668.1 | XP_027823469.1 | 7 - 8 | 1062 | 87 | ||
More Information
Set Membership: |
|

CHPT1
Genomic Map