Assay Details
| Gene Symbol: LOC101115778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005920.4 | XP_004005969.2 | 4 - 5 | 1191 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101115778| Gene Symbol: LOC101115778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005920.4 | XP_004005969.2 | 4 - 5 | 1191 | 84 | |
Set Membership: |
|