Assay Details
| Gene Symbol: PSMD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015106.3 | XP_004015155.1 | 6 - 7 | 642 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSMD7| Gene Symbol: PSMD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015106.3 | XP_004015155.1 | 6 - 7 | 642 | 64 | |
Set Membership: |
|