Assay Details
| Gene Symbol: ZFP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959134.1 | XP_027814935.1 | 1 - 2 | 1436 | 93 | |
| XM_027959135.1 | XP_027814936.1 | 2 - 3 | 639 | 93 | ||
| XM_027959136.1 | XP_027814937.1 | 3 - 4 | 596 | 93 | ||
| XM_027959137.1 | XP_027814938.1 | 2 - 3 | 549 | 93 | ||
| XM_027959138.1 | XP_027814939.1 | 2 - 3 | 868 | 93 | ||
More Information
Set Membership: |
|

ZFP57
Genomic Map