Assay Details
| Gene Symbol: GTF3C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972368.1 | XP_027828169.1 | 5 - 6 | 437 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GTF3C6| Gene Symbol: GTF3C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972368.1 | XP_027828169.1 | 5 - 6 | 437 | 79 | |
Set Membership: |
|