Assay Details
| Gene Symbol: RNF145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009029.4 | XP_004009078.1 | 4 - 5 | 730 | 72 | |
| XM_012178876.3 | XP_012034266.1 | 5 - 6 | 707 | 72 | ||
| XM_027970465.1 | XP_027826266.1 | 6 - 7 | 770 | 72 | ||
More Information
Set Membership: |
|

RNF145
Genomic Map