Assay Details
| Gene Symbol: MIA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017912.4 | XP_004017961.2 | 14 - 15 | 1320 | 83 | |
| XM_012098847.3 | XP_011954237.2 | 20 - 21 | 3143 | 83 | ||
| XM_012098849.3 | XP_011954239.2 | 19 - 20 | 3116 | 83 | ||
| XM_012098850.3 | XP_011954240.2 | 20 - 21 | 3143 | 83 | ||
| XM_012098851.3 | XP_011954241.2 | 19 - 20 | 3116 | 83 | ||
| XM_012098852.3 | XP_011954242.2 | 15 - 16 | 1269 | 83 | ||
| XM_012098853.3 | XP_011954243.2 | 15 - 16 | 2337 | 83 | ||
| XM_012098854.3 | XP_011954244.2 | 15 - 16 | 1347 | 83 | ||
| XM_012098855.3 | XP_011954245.2 | 15 - 16 | 2217 | 83 | ||
| XM_012098857.3 | XP_011954247.2 | 15 - 16 | 2208 | 83 | ||
| XM_012098858.3 | XP_011954248.2 | 14 - 15 | 1320 | 83 | ||
| XM_027957293.1 | XP_027813094.1 | 15 - 16 | 1347 | 83 | ||
| XM_027957294.1 | XP_027813095.1 | 15 - 16 | 2336 | 83 | ||
| XM_027957295.1 | XP_027813096.1 | 15 - 16 | 2208 | 83 | ||
More Information
Set Membership: |
|

MIA2
Genomic Map