Assay Details
| Gene Symbol: LOC101102169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015198.4 | XP_004015247.1 | 6 - 7 | 676 | 114 | |
| XM_012190353.3 | XP_012045743.1 | 6 - 7 | 679 | 114 | ||
| XM_027977414.1 | XP_027833215.1 | 2 - 3 | 571 | 114 | ||
More Information
Set Membership: |
|

LOC101102169
Genomic Map