Assay Details
| Gene Symbol: OCLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012096797.3 | XP_011952187.1 | 5 - 6 | 1279 | 63 | |
| XM_015101255.2 | XP_014956741.1 | 5 - 6 | 1248 | 63 | ||
| XM_015101256.2 | XP_014956742.1 | 5 - 6 | 1172 | 63 | ||
| XM_015101257.2 | XP_014956743.1 | 5 - 6 | 1180 | 63 | ||
More Information
Set Membership: |
|

OCLN
Genomic Map