Assay Details
| Gene Symbol: AP3M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021484.4 | XP_004021533.2 | 8 - 9 | 1506 | 74 | |
| XM_012105379.3 | XP_011960769.2 | 8 - 9 | 1566 | 74 | ||
More Information
Set Membership: |
|

AP3M1
Genomic Map