Assay Details
| Gene Symbol: C5H19orf53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008478.3 | XP_004008527.1 | 2 - 3 | 189 | 98 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5H19orf53| Gene Symbol: C5H19orf53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008478.3 | XP_004008527.1 | 2 - 3 | 189 | 98 | |