Assay Details
| Gene Symbol: FRZB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012131849.2 | XP_011987239.1 | 6 - 7 | 1238 | 67 | |
| XM_027964769.1 | XP_027820570.1 | 5 - 6 | 1521 | 67 | ||
More Information
Set Membership: |
|

FRZB
Genomic Map