Assay Details
| Gene Symbol: PON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007735.4 | XP_004007784.2 | 4 - 5 | 470 | 82 | |
| XM_027968471.1 | XP_027824272.1 | 4 - 5 | 453 | 82 | ||
More Information
Set Membership: |
|

PON1
Genomic Map