Assay Details
| Gene Symbol: QKI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972641.1 | XP_027828442.1 | 3 - 4 | 458 | 64 | |
| XM_027972642.1 | XP_027828443.1 | 3 - 4 | 458 | 64 | ||
| XM_027972643.1 | XP_027828444.1 | 3 - 4 | 458 | 64 | ||
| XM_027972644.1 | XP_027828445.1 | 3 - 4 | 458 | 64 | ||
| XM_027972645.1 | XP_027828446.1 | 3 - 4 | 458 | 64 | ||
More Information
Set Membership: |
|

QKI
Genomic Map