Assay Details
| Gene Symbol: HRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012190643.3 | XP_012046033.2 | 4 - 5 | 2329 | 64 | |
| XM_012190644.3 | XP_012046034.2 | 5 - 6 | 2171 | 64 | ||
More Information
Set Membership: |
|

HRC
Genomic Map