Assay Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012182462.3 | XP_012037852.2 | 11 - 12 | 1053 | 101 | |
| XM_015092270.2 | XP_014947756.2 | 10 - 11 | 2574 | 101 | ||
| XM_027975692.1 | XP_027831493.1 | 8 - 9 | 2334 | 101 | ||
More Information
Set Membership: |
|

TDRD10
Genomic Map