Assay Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012182462.3 | XP_012037852.2 | 8 - 9 | 602 | 77 | |
| XM_015092270.2 | XP_014947756.2 | 7 - 8 | 2123 | 77 | ||
| XM_027975692.1 | XP_027831493.1 | 5 - 6 | 1883 | 77 | ||
More Information
Set Membership: |
|

TDRD10
Genomic Map