Assay Details
| Gene Symbol: PUS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019523.4 | XP_004019572.1 | 2 - 3 | 488 | 68 | |
| XM_012101874.3 | XP_011957264.1 | 2 - 3 | 732 | 68 | ||
| XM_012101875.3 | XP_011957265.1 | 2 - 3 | 687 | 68 | ||
| XM_012101876.3 | XP_011957266.1 | 2 - 3 | 814 | 68 | ||
| XM_027959265.1 | XP_027815066.1 | 3 - 4 | 855 | 68 | ||
More Information
Set Membership: |
|

PUS3
Genomic Map