Assay Details
| Gene Symbol: LOC114112898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963982.1 | XP_027819783.1 | 7 - 8 | 1353 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114112898| Gene Symbol: LOC114112898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963982.1 | XP_027819783.1 | 7 - 8 | 1353 | 71 | |
Set Membership: |
|