Assay Details
| Gene Symbol: LOC101117547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960073.1 | XP_027815874.1 | 13 - 14 | 1629 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117547| Gene Symbol: LOC101117547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960073.1 | XP_027815874.1 | 13 - 14 | 1629 | 68 | |
Set Membership: |
|