Assay Details
| Gene Symbol: LOC101119694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004118.4 | XP_004004167.2 | 14 - 15 | 1925 | 104 | |
| Gene Symbol: LOC114113060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964735.1 | XP_027820536.1 | 6 - 7 | 740 | 104 | |
| Gene Symbol: LOC114113061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964736.1 | XP_027820537.1 | 3 - 4 | 299 | 104 | |
More Information
Set Membership: |
|

LOC101119694
Genomic Map