Assay Details
| Gene Symbol: LOC101119694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004118.4 | XP_004004167.2 | 8 - 9 | 1119 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101119694| Gene Symbol: LOC101119694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004118.4 | XP_004004167.2 | 8 - 9 | 1119 | 106 | |
Set Membership: |
|