Assay Details
| Gene Symbol: GLRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004453.4 | XP_004004502.2 | 7 - 8 | 940 | 82 | |
| XM_012129216.3 | XP_011984606.1 | 7 - 8 | 925 | 82 | ||
More Information
Set Membership: |
|

GLRA3
Genomic Map