Assay Details
| Gene Symbol: LOC101122306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980035.1 | XP_027835836.1 | 8 - 9 | 3504 | 72 | |
| XM_027980036.1 | XP_027835837.1 | 10 - 11 | 1770 | 72 | ||
| XM_027980037.1 | XP_027835838.1 | 8 - 9 | 3503 | 72 | ||
| XM_027980038.1 | XP_027835839.1 | 10 - 11 | 1770 | 72 | ||
More Information
Set Membership: |
|

LOC101122306
Genomic Map