Assay Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961428.1 | XP_027817229.1 | 2 - 3 | 687 | 87 | |
| XM_027961429.1 | XP_027817230.1 | 2 - 3 | 687 | 87 | ||
More Information
Set Membership: |
|

ZNF500
Genomic Map