Assay Details
| Gene Symbol: SFTPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012169361.3 | XP_012024751.1 | 7 - 8 | 835 | 75 | |
| XM_027966740.1 | XP_027822541.1 | 7 - 8 | 840 | 75 | ||
| XM_027966741.1 | XP_027822542.1 | 7 - 8 | 840 | 75 | ||
| XM_027966742.1 | XP_027822543.1 | 7 - 8 | 771 | 75 | ||
| GenBank mRNA | AF076635.1 | - | 3 - 4 | 385 | 75 | |
| AF076636.1 | - | 3 - 4 | 314 | 75 | ||
| AF107544.1 | - | 7 - 8 | 821 | 75 | ||
More Information
Set Membership: |
|

SFTPB
Genomic Map