Assay Details
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980158.1 | XP_027835959.1 | 5 - 6 | 709 | 68 | |
| XM_027980159.1 | XP_027835960.1 | 5 - 6 | 715 | 68 | ||
More Information
Set Membership: |
|

OTULIN
Genomic Map