Assay Details
| Gene Symbol: LOC101115252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961640.1 | XP_027817441.1 | 6 - 7 | 916 | 74 | |
| XM_027961641.1 | XP_027817442.1 | 6 - 7 | 727 | 74 | ||
| XM_027961642.1 | XP_027817443.1 | 7 - 8 | 779 | 74 | ||
| XM_027961643.1 | XP_027817444.1 | 6 - 7 | 669 | 74 | ||
| XM_027961644.1 | XP_027817445.1 | 5 - 6 | 708 | 74 | ||
More Information
Set Membership: |
|

LOC101115252
Genomic Map