Assay Details
| Gene Symbol: HTR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012148490.3 | XP_012003880.1 | 2 - 3 | 1550 | 69 | |
| XM_012148499.3 | XP_012003889.1 | 2 - 3 | 1189 | 69 | ||
More Information
Set Membership: |
|

HTR2B
Genomic Map