Assay Details
| Gene Symbol: TSPAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973481.1 | XP_027829282.1 | 5 - 6 | 476 | 68 | |
| XM_027973487.1 | XP_027829288.1 | 5 - 6 | 476 | 68 | ||
More Information
Set Membership: |
|

TSPAN2
Genomic Map