Assay Details
| Gene Symbol: BORCS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969354.1 | XP_027825155.1 | 3 - 4 | 846 | 66 | |
| XM_027969356.1 | XP_027825157.1 | 3 - 4 | 846 | 66 | ||
| XM_027969357.1 | XP_027825158.1 | 3 - 4 | 846 | 66 | ||
| XM_027969358.1 | XP_027825159.1 | 3 - 4 | 1289 | 66 | ||
More Information
Set Membership: |
|

BORCS8
Genomic Map