Assay Details
| Gene Symbol: C14H16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015056.4 | XP_004015105.2 | 16 - 17 | 1290 | 66 | |
| XM_027978039.1 | XP_027833840.1 | 15 - 16 | 1341 | 66 | ||
More Information
Set Membership: |
|

C14H16orf70
Genomic Map