Assay Details
| Gene Symbol: C14H16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015056.4 | XP_004015105.2 | 2 - 3 | 170 | 52 | |
| XM_027978039.1 | XP_027833840.1 | 1 - 2 | 221 | 52 | ||
More Information
Set Membership: |
|

C14H16orf70
Genomic Map