Assay Details
| Gene Symbol: C14H16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015056.4 | XP_004015105.2 | 5 - 6 | 384 | 126 | |
| XM_027978039.1 | XP_027833840.1 | 4 - 5 | 435 | 126 | ||
More Information
Set Membership: |
|

C14H16orf70
Genomic Map