Assay Details
| Gene Symbol: C14H16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015056.4 | XP_004015105.2 | 8 - 9 | 654 | 70 | |
| XM_027978039.1 | XP_027833840.1 | 7 - 8 | 705 | 70 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C14H16orf70| Gene Symbol: C14H16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015056.4 | XP_004015105.2 | 8 - 9 | 654 | 70 | |
| XM_027978039.1 | XP_027833840.1 | 7 - 8 | 705 | 70 | ||