Assay Details
| Gene Symbol: DNAJC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016366.4 | XP_004016415.3 | 2 - 3 | 181 | 79 | |
| XM_012096147.3 | XP_011951537.2 | 2 - 3 | 181 | 79 | ||
| XM_012096148.3 | XP_011951538.2 | 2 - 3 | 209 | 79 | ||
More Information
Set Membership: |
|

DNAJC24
Genomic Map