Assay Details
| Gene Symbol: PSKH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015081.4 | XP_004015130.3 | 2 - 3 | 1145 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSKH1| Gene Symbol: PSKH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015081.4 | XP_004015130.3 | 2 - 3 | 1145 | 73 | |
Set Membership: |
|