Assay Details
| Gene Symbol: OGFOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015002.3 | XP_004015051.2 | 5 - 6 | 630 | 55 | |
| XM_012189806.2 | XP_012045196.1 | 3 - 4 | 304 | 55 | ||
| XM_027977851.1 | XP_027833652.1 | 5 - 6 | 630 | 55 | ||
More Information
Set Membership: |
|

OGFOD1
Genomic Map