Assay Details
| Gene Symbol: KLK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015397.4 | XP_004015446.1 | 4 - 5 | 914 | 82 | |
| XM_027977527.1 | XP_027833328.1 | 2 - 3 | 521 | 82 | ||
More Information
Set Membership: |
|

KLK8
Genomic Map