Assay Details
| Gene Symbol: DAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018458.4 | XP_004018507.2 | 1 - 2 | 708 | 82 | |
| XM_027957633.1 | XP_027813434.1 | 2 - 3 | 526 | 82 | ||
| XM_027957634.1 | XP_027813435.1 | 2 - 3 | 443 | 82 | ||
| XM_027957635.1 | XP_027813436.1 | 2 - 3 | 537 | 82 | ||
More Information
Set Membership: |
|

DAG1
Genomic Map