Assay Details
| Gene Symbol: S100PBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012149068.3 | XP_012004458.2 | 3 - 4 | 2196 | 75 | |
| XM_012149084.3 | XP_012004474.2 | 4 - 5 | 1960 | 75 | ||
| XM_012149123.3 | XP_012004513.2 | 4 - 5 | 1210 | 75 | ||
| XM_012149133.3 | XP_012004523.2 | 4 - 5 | 1221 | 75 | ||
| XM_012149151.3 | XP_012004541.2 | 4 - 5 | 1196 | 75 | ||
| XM_012149242.3 | XP_012004632.2 | 3 - 4 | 1052 | 75 | ||
| XM_012149251.3 | XP_012004641.1 | 3 - 4 | 1426 | 75 | ||
| XM_012149302.3 | XP_012004692.1 | 3 - 4 | 302 | 75 | ||
| XM_027965337.1 | XP_027821138.1 | 3 - 4 | 2193 | 75 | ||
| XM_027965338.1 | XP_027821139.1 | 3 - 4 | 2169 | 75 | ||
| XM_027965339.1 | XP_027821140.1 | 3 - 4 | 327 | 75 | ||
More Information
Set Membership: |
|

S100PBP
Genomic Map