Assay Details
| Gene Symbol: LOC101115787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968374.1 | XP_027824175.1 | 9 - 10 | 819 | 86 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101115787| Gene Symbol: LOC101115787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968374.1 | XP_027824175.1 | 9 - 10 | 819 | 86 | |