Assay Details
| Gene Symbol: LOC101109474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100011.2 | XP_014955497.2 | 8 - 9 | 1011 | 87 | |
| XM_015100014.2 | XP_014955500.2 | 6 - 7 | 688 | 87 | ||
| XM_015100015.2 | XP_014955501.2 | 2 - 3 | 215 | 87 | ||
| XM_027978441.1 | XP_027834242.1 | 8 - 9 | 922 | 87 | ||
| XM_027978442.1 | XP_027834243.1 | 6 - 7 | 1077 | 87 | ||
| XM_027978443.1 | XP_027834244.1 | 7 - 8 | 815 | 87 | ||
| XM_027978444.1 | XP_027834245.1 | 7 - 8 | 716 | 87 | ||
| XM_027978445.1 | XP_027834246.1 | 6 - 7 | 602 | 87 | ||
| XM_027978446.1 | XP_027834247.1 | 6 - 7 | 646 | 87 | ||
| XM_027978447.1 | XP_027834248.1 | 6 - 7 | 617 | 87 | ||
More Information
Set Membership: |
|

LOC101109474
Genomic Map