Assay Details
| Gene Symbol: GOT1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021775.4 | XP_004021824.1 | 7 - 8 | 1462 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GOT1L1| Gene Symbol: GOT1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021775.4 | XP_004021824.1 | 7 - 8 | 1462 | 62 | |
Set Membership: |
|