Assay Details
| Gene Symbol: PAQR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971529.1 | XP_027827330.1 | 4 - 5 | 600 | 60 | |
| XM_027971530.1 | XP_027827331.1 | 4 - 5 | 1681 | 60 | ||
More Information
Set Membership: |
|

PAQR5
Genomic Map