Assay Details
| Gene Symbol: PKLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976116.1 | XP_027831917.1 | 9 - 10 | 1519 | 63 | |
| XM_027976118.1 | XP_027831919.1 | 10 - 11 | 1518 | 63 | ||
More Information
Set Membership: |
|

PKLR
Genomic Map