Assay Details
| Gene Symbol: LOC101123372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177962.3 | XP_012033352.1 | 9 - 10 | 1147 | 100 | |
| XM_012177964.3 | XP_012033354.1 | 9 - 10 | 892 | 100 | ||
More Information
Set Membership: |
|

LOC101123372
Genomic Map